Assay Details
| Gene Symbol: CKMT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204182.2 | NP_989513.2 | 2 - 3 | 407 | 74 | |
| GenBank mRNA | BU295295.1 | - | 2 - 3 | 262 | 74 | |
| CO421284.1 | - | 2 - 3 | 415 | 74 | ||
More Information
Set Membership: |
|

CKMT1B
Genomic Map