Assay Details
| Gene Symbol: CKMT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204182.2 | NP_989513.2 | 7 - 8 | 1075 | 71 | |
| GenBank mRNA | BU118323.1 | - | 3 - 4 | 321 | 71 | |
| BU339387.1 | - | 2 - 3 | 251 | 71 | ||
More Information
Set Membership: |
|

CKMT1B
Genomic Map