Assay Details
| Gene Symbol: FABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204192.3 | NP_989523.1 | 2 - 3 | 280 | 84 | |
| GenBank mRNA | AF380999.1 | - | 2 - 3 | 285 | 84 | |
| AY321365.1 | - | 2 - 3 | 257 | 84 | ||
| EU049889.1 | - | 2 - 3 | 239 | 84 | ||
More Information
Set Membership: |
|

FABP1
Genomic Map