Assay Details
| Gene Symbol: DIAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204210.1 | NP_989541.1 | 8 - 9 | 1003 | 63 | |
| XM_015278156.2 | XP_015133642.2 | 9 - 10 | 1073 | 63 | ||
| XM_015278157.2 | XP_015133643.2 | 9 - 10 | 1074 | 63 | ||
| XM_015278159.2 | XP_015133645.2 | 8 - 9 | 1040 | 63 | ||
| XM_015278160.2 | XP_015133646.2 | 9 - 10 | 1074 | 63 | ||
| XM_015278161.2 | XP_015133647.2 | 7 - 8 | 1019 | 63 | ||
| XM_015278162.2 | XP_015133648.2 | 9 - 10 | 1074 | 63 | ||
| XM_025149458.1 | XP_025005226.1 | 8 - 9 | 1052 | 63 | ||
| GenBank mRNA | AB025226.1 | - | 8 - 9 | 1003 | 63 | |
More Information
Set Membership: |
|

DIAPH2
Genomic Map