Assay Details
| Gene Symbol: CCND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204213.1 | NP_989544.1 | 3 - 4 | 569 | 66 | |
| XM_015292118.2 | XP_015147604.1 | 4 - 5 | 703 | 66 | ||
| XM_025149632.1 | XP_025005400.1 | 4 - 5 | 703 | 66 | ||
More Information
Set Membership: |
|

CCND2
Genomic Map