Assay Details
| Gene Symbol: MYOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204214.2 | NP_989545.2 | 1 - 2 | 716 | 87 | |
| GenBank mRNA | FJ977569.1 | - | 1 - 2 | 694 | 87 | |
| X16189.1 | - | 713 | 87 | |||
More Information
Set Membership: |
|

MYOD1
Genomic Map