Assay Details
| Gene Symbol: RPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204217.1 | NP_989548.1 | 4 - 5 | 364 | 102 | |
| GenBank mRNA | AY215074.1 | - | 4 - 5 | 364 | 102 | |
| BX930857.2 | - | 4 - 5 | 351 | 102 | ||
| BX933819.1 | - | 586 | 102 | |||
| BX935295.2 | - | 1 - 2 | 713 | 102 | ||
| CR352568.1 | - | 1 - 2 | 707 | 102 | ||
| X07257.1 | - | 309 | 102 | |||
More Information
Set Membership: |
|

RPS17
Genomic Map