Assay Details
| Gene Symbol: DAZL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204218.1 | NP_989549.1 | 7 - 8 | 739 | 72 | |
| XM_015281256.2 | XP_015136742.1 | 7 - 8 | 1322 | 72 | ||
| XM_015281259.2 | XP_015136745.1 | 7 - 8 | 1303 | 72 | ||
| GenBank mRNA | AY211387.1 | - | 7 - 8 | 739 | 72 | |
More Information
Set Membership: |
|

DAZL
Genomic Map