Assay Details
| Gene Symbol: LITAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204267.1 | NP_989598.1 | 4 - 5 | 514 | 64 | |
| XM_015294117.2 | XP_015149603.1 | 3 - 4 | 394 | 64 | ||
| XM_015294119.2 | XP_015149605.1 | 4 - 5 | 572 | 64 | ||
| XM_015294120.2 | XP_015149606.1 | 4 - 5 | 645 | 64 | ||
| XM_015294121.2 | XP_015149607.1 | 4 - 5 | 791 | 64 | ||
| XM_015294124.2 | XP_015149610.1 | 3 - 4 | 380 | 64 | ||
| GenBank mRNA | AB058634.1 | - | 4 - 5 | 514 | 64 | |
| GU230788.1 | - | 1 - 2 | 102 | 64 | ||
More Information
Set Membership: |
|

LITAF
Genomic Map