Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204268.2 | NP_989599.1 | 1 - 2 | 827 | 61 | |
| XM_015289084.2 | XP_015144570.1 | 2 - 3 | 997 | 61 | ||
| XM_015289085.2 | XP_015144571.1 | 2 - 3 | 1141 | 61 | ||
| XM_015289086.2 | XP_015144572.1 | 2 - 3 | 1003 | 61 | ||
| XM_025152071.1 | XP_025007839.1 | 2 - 3 | 6737 | 61 | ||
| GenBank mRNA | AY035386.1 | - | 1 - 2 | 827 | 61 | |
More Information
Set Membership: |
|

NAB1
Genomic Map