Assay Details
| Gene Symbol: PAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204269.1 | NP_989600.1 | 1 - 2 | 104 | 111 | |
| XM_025153298.1 | XP_025009066.1 | 1 - 2 | 337 | 111 | ||
| XM_025153299.1 | XP_025009067.1 | 2 - 3 | 680 | 111 | ||
| XM_025153300.1 | XP_025009068.1 | 1 - 2 | 2537 | 111 | ||
| GenBank mRNA | AB080581.1 | - | 1 - 2 | 104 | 111 | |
More Information
Set Membership: |
|

PAX3
Genomic Map