Assay Details
| Gene Symbol: RPL39L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204272.1 | NP_989603.1 | 2 - 3 | 136 | 76 | |
| GenBank mRNA | AB046397.1 | - | 2 - 3 | 136 | 76 | |
| CR385515.1 | - | 1 - 2 | 374 | 76 | ||
More Information
Set Membership: |
|

RPL39L
Genomic Map