Assay Details
| Gene Symbol: NOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204274.1 | NP_989605.1 | 2 - 3 | 442 | 97 | |
| GenBank mRNA | AF463517.1 | - | 2 - 3 | 349 | 97 | |
| AJ298335.1 | - | 2 - 3 | 442 | 97 | ||
More Information
Set Membership: |
|

NOV
Genomic Map