Assay Details
| Gene Symbol: ERG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204280.1 | NP_989611.1 | 6 - 7 | 805 | 64 | |
| XM_015300250.2 | XP_015155736.1 | 7 - 8 | 1049 | 64 | ||
| XM_015300252.2 | XP_015155738.1 | 7 - 8 | 901 | 64 | ||
| XM_015300256.2 | XP_015155742.1 | 6 - 7 | 973 | 64 | ||
| XM_015300260.2 | XP_015155746.1 | 5 - 6 | 753 | 64 | ||
| XM_025150666.1 | XP_025006434.1 | 6 - 7 | 1035 | 64 | ||
| GenBank mRNA | AY065661.1 | - | 5 - 6 | 698 | 64 | |
| X77159.1 | - | 805 | 64 | |||
More Information
Set Membership: |
|

ERG
Genomic Map