Assay Details
| Gene Symbol: SOX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204281.1 | NP_989612.1 | 2 - 3 | 802 | 104 | |
| GenBank mRNA | AB012236.1 | - | 2 - 3 | 802 | 104 | |
| U12533.1 | - | 2 - 3 | 694 | 104 | ||
More Information
Set Membership: |
|

SOX9
Genomic Map