Assay Details
| Gene Symbol: HSP90B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204289.1 | NP_989620.1 | 5 - 6 | 842 | 65 | |
| GenBank mRNA | EU828792.1 | - | 5 - 6 | 741 | 65 | |
| M14772.1 | - | 5 - 6 | 842 | 65 | ||
More Information
Set Membership: |
|

HSP90B1
Genomic Map