Assay Details
| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204290.1 | NP_989621.1 | 3 - 4 | 380 | 112 | |
| GenBank mRNA | AF432506.1 | - | 3 - 4 | 380 | 112 | |
| KT165375.1 | - | 3 - 4 | 353 | 112 | ||
More Information
Set Membership: |
|

FABP4
Genomic Map