Assay Details
| Gene Symbol: UFD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204301.1 | NP_989632.1 | 3 - 4 | 303 | 61 | |
| GenBank mRNA | AF228284.1 | - | 3 - 4 | 303 | 61 | |
| AF407671.1 | - | 4 - 5 | 319 | 61 | ||
More Information
Set Membership: |
|

UFD1L
Genomic Map