Assay Details
| Gene Symbol: MAGOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308098.1 | NP_001295027.1 | 1 - 2 | 129 | 73 | |
| GenBank mRNA | AF383945.1 | - | 1 - 2 | 110 | 73 | |
| BU106550.1 | - | 1 - 2 | 103 | 73 | ||
| BU285565.1 | - | 1 - 2 | 134 | 73 | ||
More Information
Set Membership: |
|

MAGOH
Genomic Map