Assay Details
| Gene Symbol: WNT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204336.2 | NP_989667.2 | 964 | 76 | ||
| GenBank mRNA | AF182403.1 | - | 714 | 76 | ||
| AF346628.1 | - | 924 | 76 | |||
| BX935899.1 | - | 965 | 76 | |||
More Information
Set Membership: |
|

WNT2B
Genomic Map