Assay Details
| Gene Symbol: LSP1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303036.1 | NP_001289965.1 | 7 - 8 | 837 | 67 | |
| XM_015286521.1 | XP_015142007.1 | 7 - 8 | 1074 | 67 | ||
| XM_015286522.1 | XP_015142008.1 | 7 - 8 | 1074 | 67 | ||
| XM_025150540.1 | XP_025006308.1 | 7 - 8 | 863 | 67 | ||
| XM_025150541.1 | XP_025006309.1 | 8 - 9 | 954 | 67 | ||
| GenBank mRNA | AB101641.1 | - | 6 - 7 | 657 | 67 | |
| AJ851520.1 | - | 7 - 8 | 837 | 67 | ||
More Information
Set Membership: |
|

LSP1P1
Genomic Map