Assay Details
| Gene Symbol: TP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204351.1 | NP_989682.1 | 5 - 6 | 710 | 64 | |
| XM_015291361.2 | XP_015146847.1 | 7 - 8 | 1071 | 64 | ||
| XM_015291362.2 | XP_015146848.1 | 7 - 8 | 1057 | 64 | ||
| XM_015291363.2 | XP_015146849.1 | 5 - 6 | 855 | 64 | ||
| XM_015291364.2 | XP_015146850.1 | 5 - 6 | 781 | 64 | ||
| XM_015291366.2 | XP_015146852.1 | 7 - 8 | 1058 | 64 | ||
| XM_015291367.2 | XP_015146853.1 | 7 - 8 | 1063 | 64 | ||
| GenBank mRNA | AB045224.1 | - | 5 - 6 | 710 | 64 | |
| CR388649.1 | - | 5 - 6 | 822 | 64 | ||
| KU981085.1 | - | 7 - 8 | 1076 | 64 | ||
| KU981086.1 | - | 5 - 6 | 846 | 64 | ||
| MH238464.1 | - | 7 - 8 | 1076 | 64 | ||
| MH238465.1 | - | 5 - 6 | 846 | 64 | ||
More Information
Set Membership: |
|

TP63
Genomic Map