Assay Details
| Gene Symbol: NPFFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204362.1 | NP_989693.1 | 3 - 4 | 462 | 93 | |
| GenBank mRNA | AB120326.1 | - | 3 - 4 | 462 | 93 | |
More Information
Set Membership: |
|

NPFFR1
Genomic Map