Assay Details
| Gene Symbol: NPVF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204363.1 | NP_989694.1 | 2 - 3 | 554 | 120 | |
| GenBank mRNA | AB120325.1 | - | 2 - 3 | 554 | 120 | |
| CR406155.1 | - | 2 - 3 | 531 | 120 | ||
More Information
Set Membership: |
|

NPVF
Genomic Map