Assay Details
| Gene Symbol: TRMT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204368.1 | NP_989699.1 | 1 - 2 | 134 | 58 | |
| GenBank mRNA | AY139090.1 | - | 1 - 2 | 134 | 58 | |
More Information
Set Membership: |
|

TRMT11
Genomic Map