Assay Details
| Gene Symbol: ARSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204372.1 | NP_989703.1 | 5 - 6 | 525 | 76 | |
| GenBank mRNA | AF534911.1 | - | 5 - 6 | 525 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARSH| Gene Symbol: ARSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204372.1 | NP_989703.1 | 5 - 6 | 525 | 76 | |
| GenBank mRNA | AF534911.1 | - | 5 - 6 | 525 | 76 | |
Set Membership: |
|