Assay Details
| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204382.1 | NP_989713.1 | 5 - 6 | 784 | 65 | |
| XM_025152732.1 | XP_025008500.1 | 5 - 6 | 455 | 65 | ||
| GenBank mRNA | AB095027.2 | - | 5 - 6 | 784 | 65 | |
More Information
Set Membership: |
|

DDAH1
Genomic Map