Assay Details
| Gene Symbol: SLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204391.1 | NP_989722.1 | 4 - 5 | 357 | 70 | |
| XM_015283077.2 | XP_015138563.1 | 5 - 6 | 559 | 70 | ||
| XM_015283079.2 | XP_015138565.1 | 5 - 6 | 544 | 70 | ||
| XM_025147006.1 | XP_025002774.1 | 4 - 5 | 522 | 70 | ||
| GenBank mRNA | AJ721107.1 | - | 4 - 5 | 541 | 70 | |
| AY278230.1 | - | 4 - 5 | 357 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025147006.1 transcripts.
Set Membership: |
|

SLA
Genomic Map