Assay Details
| Gene Symbol: LHFPL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204398.1 | NP_989729.1 | 756 | 73 | ||
| XM_015298680.2 | XP_015154166.2 | 527 | 73 | |||
| GenBank mRNA | AY265159.1 | - | 756 | 73 | ||
More Information
Set Membership: |
|

LHFPL5
Genomic Map