Assay Details
| Gene Symbol: RIPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204402.2 | NP_989733.2 | 8 - 9 | 1140 | 68 | |
| XM_015275819.2 | XP_015131305.1 | 8 - 9 | 1309 | 68 | ||
| XM_025147007.1 | XP_025002775.1 | 8 - 9 | 1531 | 68 | ||
| GenBank mRNA | AB108485.1 | - | 8 - 9 | 1103 | 68 | |
| AJ851558.1 | - | 8 - 9 | 1135 | 68 | ||
More Information
Set Membership: |
|

RIPK1
Genomic Map