Assay Details
| Gene Symbol: TNFSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204409.1 | NP_989740.1 | 1 - 2 | 333 | 67 | |
| GenBank mRNA | AB105911.2 | - | 1 - 2 | 373 | 67 | |
More Information
Set Membership: |
|

TNFSF8
Genomic Map