Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204410.1 | NP_989741.1 | 9 - 10 | 911 | 65 | |
| GenBank mRNA | L24906.1 | - | 9 - 10 | 911 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPARC| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204410.1 | NP_989741.1 | 9 - 10 | 911 | 65 | |
| GenBank mRNA | L24906.1 | - | 9 - 10 | 911 | 65 | |
Set Membership: |
|