Assay Details
| Gene Symbol: GRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204411.1 | NP_989742.1 | 4 - 5 | 472 | 88 | |
| XM_015295374.2 | XP_015150860.1 | 6 - 7 | 5375 | 88 | ||
| XM_015295375.2 | XP_015150861.1 | 6 - 7 | 843 | 88 | ||
| XM_015295376.2 | XP_015150862.1 | 6 - 7 | 721 | 88 | ||
| XM_015295378.2 | XP_015150864.1 | 5 - 6 | 5309 | 88 | ||
| XM_025141539.1 | XP_024997307.1 | 6 - 7 | 851 | 88 | ||
| XM_025141540.1 | XP_024997308.1 | 6 - 7 | 720 | 88 | ||
| GenBank mRNA | L19258.1 | - | 4 - 5 | 472 | 88 | |
More Information
Set Membership: |
|

GRB2
Genomic Map