Assay Details
| Gene Symbol: ALG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204435.1 | NP_989766.1 | 2 - 3 | 166 | 80 | |
| XM_015290939.2 | XP_015146425.1 | 2 - 3 | 765 | 80 | ||
| XM_025152743.1 | XP_025008511.1 | 2 - 3 | 491 | 80 | ||
| XM_025152744.1 | XP_025008512.1 | 2 - 3 | 186 | 80 | ||
| XM_025152745.1 | XP_025008513.1 | 2 - 3 | 134 | 80 | ||
| GenBank mRNA | AJ535824.1 | - | 2 - 3 | 166 | 80 | |
| AJ719772.1 | - | 2 - 3 | 148 | 80 | ||
More Information
Set Membership: |
|

ALG6
Genomic Map