Assay Details
| Gene Symbol: TBX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204453.4 | NP_989784.2 | 5 - 6 | 883 | 77 | |
| GenBank mRNA | AY173127.2 | - | 5 - 6 | 883 | 77 | |
| AY550994.1 | - | 2 - 3 | 293 | 77 | ||
More Information
Set Membership: |
|

TBX18
Genomic Map