Assay Details
| Gene Symbol: CRHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204454.1 | NP_989785.1 | 6 - 7 | 694 | 66 | |
| XM_015281045.2 | XP_015136531.1 | 7 - 8 | 851 | 66 | ||
| XM_015281046.2 | XP_015136532.1 | 3 - 4 | 336 | 66 | ||
| GenBank mRNA | AJ557031.2 | - | 6 - 7 | 694 | 66 | |
| KY196000.1 | - | 7 - 8 | 739 | 66 | ||
More Information
Set Membership: |
|

CRHR2
Genomic Map