Assay Details
| Gene Symbol: KCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204463.1 | NP_989794.1 | 1443 | 103 | ||
| GenBank mRNA | AY329361.1 | - | 1443 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNA2| Gene Symbol: KCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204463.1 | NP_989794.1 | 1443 | 103 | ||
| GenBank mRNA | AY329361.1 | - | 1443 | 103 | ||
Set Membership: |
|