Assay Details
| Gene Symbol: CCNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204470.2 | NP_989801.2 | 8 - 9 | 1053 | 129 | |
| XM_025142479.1 | XP_024998247.1 | 4 - 5 | 2071 | 129 | ||
| XM_025142480.1 | XP_024998248.1 | 4 - 5 | 404 | 129 | ||
| XM_025142481.1 | XP_024998249.1 | 4 - 5 | 417 | 129 | ||
| GenBank mRNA | AJ720385.1 | - | 8 - 9 | 1053 | 129 | |
| AY333181.1 | - | 3 - 4 | 462 | 129 | ||
More Information
Set Membership: |
|

CCNL2
Genomic Map