Assay Details
| Gene Symbol: HS6ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204482.1 | NP_989813.1 | 1 - 2 | 764 | 101 | |
| GenBank mRNA | AB093515.1 | - | 1 - 2 | 764 | 101 | |
| BX935211.2 | - | 1 - 2 | 559 | 101 | ||
| CR386631.1 | - | 1 - 2 | 561 | 101 | ||
More Information
Set Membership: |
|

HS6ST1
Genomic Map