Assay Details
| Gene Symbol: LOC395159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204498.2 | NP_989829.1 | 2 - 3 | 417 | 95 | |
| GenBank mRNA | AY390351.1 | - | 2 - 3 | 361 | 95 | |
| BU340855.1 | - | 2 - 3 | 431 | 95 | ||
| BU353132.1 | - | 1 - 2 | 58 | 95 | ||
| BU401663.1 | - | 1 - 2 | 56 | 95 | ||
| CK987019.1 | - | 2 - 3 | 358 | 95 | ||
More Information
Set Membership: |
|

LOC395159
Genomic Map