Assay Details
| Gene Symbol: LOC395159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204498.2 | NP_989829.1 | 4 - 5 | 607 | 99 | |
| GenBank mRNA | AY390351.1 | - | 4 - 5 | 551 | 99 | |
| BU340855.1 | - | 4 - 5 | 621 | 99 | ||
| BU353132.1 | - | 3 - 4 | 248 | 99 | ||
| BU401663.1 | - | 3 - 4 | 246 | 99 | ||
More Information
Set Membership: |
|

LOC395159
Genomic Map