Assay Details
| Gene Symbol: IL17RD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204515.1 | NP_989846.1 | 2 - 3 | 207 | 78 | |
| XM_025154411.1 | XP_025010179.1 | 2 - 3 | 108 | 78 | ||
| XM_025154412.1 | XP_025010180.1 | 2 - 3 | 287 | 78 | ||
| GenBank mRNA | AJ508679.1 | - | 1 - 2 | 63 | 78 | |
| AY278204.1 | - | 2 - 3 | 207 | 78 | ||
| CR386972.1 | - | 2 - 3 | 104 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025154411.1 transcripts.
Set Membership: |
|

IL17RD
Genomic Map