Assay Details
| Gene Symbol: P2RX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204519.1 | NP_989850.1 | 10 - 11 | 1122 | 58 | |
| XM_015295662.2 | XP_015151148.1 | 12 - 13 | 1385 | 58 | ||
| GenBank mRNA | AJ511278.1 | - | 10 - 11 | 1122 | 58 | |
More Information
Set Membership: |
|

P2RX1
Genomic Map