Assay Details
| Gene Symbol: SH3GL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204530.1 | NP_989861.1 | 1 - 2 | 63 | 65 | |
| GenBank mRNA | AJ439350.1 | - | 1 - 2 | 63 | 65 | |
| BX934239.1 | - | 1 - 2 | 42 | 65 | ||
More Information
Set Membership: |
|

SH3GL2
Genomic Map