Assay Details
| Gene Symbol: ICOSLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205084.2 | NP_990415.1 | 2 - 3 | 115 | 83 | |
| GenBank mRNA | BU245144.1 | - | 2 - 3 | 115 | 83 | |
| Y08823.1 | - | 102 | 83 | |||
More Information
Set Membership: |
|

ICOSLG
Genomic Map