Assay Details
| Gene Symbol: ICOSLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205084.2 | NP_990415.1 | 5 - 6 | 949 | 93 | |
| GenBank mRNA | Y08823.1 | - | 936 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ICOSLG| Gene Symbol: ICOSLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205084.2 | NP_990415.1 | 5 - 6 | 949 | 93 | |
| GenBank mRNA | Y08823.1 | - | 936 | 93 | ||
Set Membership: |
|