Assay Details
| Gene Symbol: GFI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205091.1 | NP_990422.1 | 5 - 6 | 752 | 59 | |
| GenBank mRNA | Y10898.1 | - | 752 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GFI1B| Gene Symbol: GFI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205091.1 | NP_990422.1 | 5 - 6 | 752 | 59 | |
| GenBank mRNA | Y10898.1 | - | 752 | 59 | ||
Set Membership: |
|