Assay Details
| Gene Symbol: MAPK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205095.1 | NP_990426.1 | 4 - 5 | 383 | 74 | |
| GenBank mRNA | AB000807.1 | - | 4 - 5 | 383 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAPK9| Gene Symbol: MAPK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205095.1 | NP_990426.1 | 4 - 5 | 383 | 74 | |
| GenBank mRNA | AB000807.1 | - | 4 - 5 | 383 | 74 | |
Set Membership: |
|