Assay Details
Gene Symbol: CRTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205100.1 | NP_990431.1 | 2 - 3 | 629 | 63 | |
XM_015281303.2 | XP_015136789.1 | 2 - 3 | 653 | 63 | ||
GenBank mRNA | X97607.1 | - | 629 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
