Assay Details
| Gene Symbol: TBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205103.1 | NP_990434.1 | 2 - 3 | 443 | 61 | |
| XM_015284047.1 | XP_015139533.1 | 3 - 4 | 658 | 61 | ||
| XM_025148547.1 | XP_025004315.1 | 3 - 4 | 655 | 61 | ||
| GenBank mRNA | AF221563.1 | - | 1 - 2 | 347 | 61 | |
| AF221564.1 | - | 1 - 2 | 347 | 61 | ||
| D83126.1 | - | 3 - 4 | 619 | 61 | ||
| D83127.1 | - | 2 - 3 | 443 | 61 | ||
| EU334508.1 | - | 2 - 3 | 401 | 61 | ||
More Information
Set Membership: |
|

TBP
Genomic Map